4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Synonyms: 4-Benzyloxy-2,3-difluorobenzeneboronic acid pinacol ester, 4-(Benzyloxy)-2,3-difluorophenylboronic acid pinacol ester

31273-500MG 31273-1G 31273-5G 31273-25G
77325-236EA 99.91 USD
77325-236 77342-768 77361-590 77373-252
4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester ≥97%
4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₁BF₂O₃
MDL Number: MFCD23098821
CAS Number: 2121515-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR