1-Bromo-3-fluoro-2-methyl-4-(trifluoromethyl)benzene ≥95%
Supplier: AOBChem USA
31253-500MG
31253-1G
31253-5G
31253-25G
77325-232EA
184.37
USD
77325-232
77347-184
77361-586
77384-728
1-Bromo-3-fluoro-2-methyl-4-(trifluoromethyl)benzene ≥95%
1-Bromo-3-fluoro-2-methyl-4-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₅BrF₄ MW: 257.023 g/mol |
MDL Number:
MFCD31714151 CAS Number: 1628524-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...