4,5-Dichloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31233-500MG 31233-1G 31233-5G
77325-230EA 326.51 USD
77325-230 77348-978 77373-250
4,5-Dichloro-2-fluorobenzyl alcohol ≥97%
4,5-Dichloro-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28786035
CAS Number: 1806298-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR