2,6-Dichloro-3-fluoroanisole ≥95%

Supplier: AOBChem USA

Warning

31213-500MG 31213-1G 31213-5G
77336-612EA 390.37 USD
77336-612 77348-974 77373-244
2,6-Dichloro-3-fluoroanisole ≥95%
2,6-Dichloro-3-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
MW: 195.018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28785947
CAS Number: 1803784-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR