3,5-Dichloro-2-fluorophenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31211-500MG 31211-1G 31211-5G 31211-25G
77325-226EA 160.68 USD
77325-226 77345-766 77361-580 77384-724
3,5-Dichloro-2-fluorophenylboronic acid pinacol ester ≥95%
3,5-Dichloro-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BCl₂FO₂
MW: 290.954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088282
CAS Number: 2121515-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR