4-Benzyloxy-5-chloro-2-fluorophenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31207-500MG 31207-1G 31207-5G 31207-25G
77325-224EA 284.28 USD
77325-224 77348-972 77361-578 77384-722
4-Benzyloxy-5-chloro-2-fluorophenylboronic acid pinacol ester ≥95%
4-Benzyloxy-5-chloro-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₁BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD29088281
CAS Number: 2121514-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR