(5-Chloro-2-fluoro-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

31192-500MG 31192-1G 31192-5G 31192-25G
77325-218EA 263.68 USD
77325-218 77348-966 77361-572 77384-716
(5-Chloro-2-fluoro-4-methoxyphenyl)methanol ≥97%
(5-Chloro-2-fluoro-4-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18441711
CAS Number: 1266177-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR