2,5-Difluoro-4-ethoxyphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31185-500MG 31185-1G 31185-5G 31185-25G
77325-216EA 141.11 USD
77325-216 77344-784 77361-570 77384-714
2,5-Difluoro-4-ethoxyphenylboronic acid pinacol ester ≥95%
2,5-Difluoro-4-ethoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BF₂O₃
MW: 284.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503400
CAS Number: 2121512-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR