3-Chloro-6-fluoro-2-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31175-500MG 31175-1G 31175-5G 31175-25G
77325-214EA 98.88 USD
77325-214 77342-686 77361-568 77373-242
3-Chloro-6-fluoro-2-methoxyphenylboronic acid pinacol ester ≥97%
3-Chloro-6-fluoro-2-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD22415054
CAS Number: 2121513-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR