2,5-Dimethyl-4-hydroxymethylphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
31159-500MG
31159-1G
31159-5G
31159-25G
77325-212EA
284.28
USD
77325-212
77348-964
77361-566
77384-712
2,5-Dimethyl-4-hydroxymethylphenylboronic acid pinacol ester ≥97%
2,5-Dimethyl-4-hydroxymethylphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₃BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD29088270 CAS Number: 2121514-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...