3-Isoproproxy-4-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31109-500MG 31109-1G 31109-5G 31109-25G
77325-208EA 123.6 USD
77325-208 77344-028 77361-560 77373-238
3-Isoproproxy-4-methylphenylboronic acid pinacol ester ≥97%
3-Isoproproxy-4-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₃
MW: 276.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088260
CAS Number: 2121514-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR