2-Bromo-6-chlorophenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Bromo-6-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

31107-1G 31107-5G 31107-25G
77340-630EA 60.77 USD
77340-630 77361-558 77373-236
2-Bromo-6-chlorophenylboronic acid pinacol ester ≥95%
2-Bromo-6-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BBrClO₂
MW: 317.414 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092541
CAS Number: 2121512-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR