2-Bromo-6-fluoro-3-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31105-1G 31105-5G 31105-25G
77341-126EA 72.1 USD
77341-126 77361-556 77373-234
2-Bromo-6-fluoro-3-methylphenylboronic acid pinacol ester ≥97%
2-Bromo-6-fluoro-3-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MW: 314.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141548
CAS Number: 2121512-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR