2-Bromo-3-trifluoromethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31097-500MG 31097-1G 31097-5G 31097-25G
77325-206EA 123.6 USD
77325-206 77344-026 77361-554 77373-232
2-Bromo-3-trifluoromethylphenylboronic acid pinacol ester ≥97%
2-Bromo-3-trifluoromethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BBrF₃O₂
MW: 350.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088258
CAS Number: 2121513-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR