2-(4-Bromonaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

31091-1G 31091-5G 31091-25G
77340-628EA 60.77 USD
77340-628 77361-548 77373-226
2-(4-Bromonaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Bromonaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₁₈BBrO₂
MW: 333.028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996405
CAS Number: 1404070-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR