6-Fluoro-2-(methylsulfanyl)phenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
31028-500MG
31028-1G
31028-5G
31028-25G
77325-202EA
141.11
USD
77325-202
77344-782
77361-542
77384-708
6-Fluoro-2-(methylsulfanyl)phenylboronic acid pinacol ester ≥97%
6-Fluoro-2-(methylsulfanyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BFO₂S Storage Temperature: Ambient |
MDL Number:
MFCD29088123 CAS Number: 2121513-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...