3-Chloro-4-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31025-500MG 31025-1G 31025-5G 31025-25G
77325-200EA 123.6 USD
77325-200 77344-024 77361-540 77373-220
3-Chloro-4-(methylthio)phenylboronic acid pinacol ester ≥97%
3-Chloro-4-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088251
CAS Number: 1438262-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR