2-Fluoro-3-(methylsulfanyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31024-500MG 31024-1G 31024-5G 31024-25G
77325-198EA 145.23 USD
77325-198 77345-122 77361-538 77384-706
2-Fluoro-3-(methylsulfanyl)phenylboronic acid pinacol ester ≥97%
2-Fluoro-3-(methylsulfanyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088250
CAS Number: 2121512-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR