Methyl 4-chloro-3-ethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

31003-500MG 31003-1G 31003-5G 31003-25G
77325-196EA 197.76 USD
77325-196 77348-958 77361-536 77384-704
Methyl 4-chloro-3-ethylbenzoate ≥95%
Methyl 4-chloro-3-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439984
CAS Number: 1345037-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR