4-Fluoro-3-(methylthio)phenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
30999-500MG
30999-1G
30999-5G
30999-25G
77325-194EA
123.6
USD
77325-194
77344-022
77361-534
77373-216
4-Fluoro-3-(methylthio)phenylboronic acid pinacol ester ≥97%
4-Fluoro-3-(methylthio)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BFO₂S MW: 268.155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29088245 CAS Number: 2121513-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...