2-Fluoro-5-(methylthio)phenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

30980-500MG 30980-1G 30980-5G 30980-25G
77325-192EA 181.28 USD
77325-192 77346-802 77361-532 77384-702
2-Fluoro-5-(methylthio)phenylboronic acid pinacol ester ≥95%
2-Fluoro-5-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088244
CAS Number: 2121512-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR