4-Ethoxymethyl-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

30972-500MG 30972-1G 30972-5G 30972-25G
77325-190EA 95.79 USD
77325-190 77342-602 77361-530 77373-214
4-Ethoxymethyl-benzoic acid ≥97%
4-Ethoxymethyl-benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00228186
CAS Number: 146781-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR