1-Bromo-4-(ethoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

30971-1G 30971-5G 30971-25G
77341-222EA 77.25 USD
77341-222 77361-528 77373-212
1-Bromo-4-(ethoxymethyl)benzene ≥97%
1-Bromo-4-(ethoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD14686973
CAS Number: 95068-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR