3-Fluoro-2-methyl-6-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30961-500MG 30961-1G 30961-5G
77336-332EA 387.28 USD
77336-332 77348-956 77373-210
3-Fluoro-2-methyl-6-(methylthio)phenylboronic acid pinacol ester ≥97%
3-Fluoro-2-methyl-6-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088242
CAS Number: 2121512-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR