2-Bromo-6-fluoro-3-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

30957-500MG 30957-1G 30957-5G 30957-25G
77325-186EA 156.56 USD
77325-186 77345-604 77361-524 77384-698
2-Bromo-6-fluoro-3-(methylsulfanyl)benzoic acid ≥97%
2-Bromo-6-fluoro-3-(methylsulfanyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD22490216
CAS Number: 1824583-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR