(2-Bromo-6-fluoro-3-methylsulfanylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

30956-500MG 30956-1G 30956-5G 30956-25G
77325-184EA 181.28 USD
77325-184 77346-798 77361-522 77384-696
(2-Bromo-6-fluoro-3-methylsulfanylphenyl)methanol ≥97%
(2-Bromo-6-fluoro-3-methylsulfanylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
MW: 251.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088238
CAS Number: 1879026-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR