2-Bromo-4-fluoro-3-methoxythioanisole ≥97%

Supplier: AOBChem USA

Warning

30954-500MG 30954-1G 30954-5G 30954-25G
77325-182EA 243.08 USD
77325-182 77348-952 77361-520 77384-694
2-Bromo-4-fluoro-3-methoxythioanisole ≥97%
2-Bromo-4-fluoro-3-methoxythioanisole

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
Storage Temperature: Ambient
MDL Number: MFCD29088236
CAS Number: 1879026-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR