2-Bromo-6-fluoro-3-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

30953-500MG 30953-1G 30953-5G 30953-25G
77325-180EA 201.88 USD
77325-180 77348-950 77361-518 77384-692
2-Bromo-6-fluoro-3-(methylthio)phenol ≥97%
2-Bromo-6-fluoro-3-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆BrFOS
Storage Temperature: Ambient
MDL Number: MFCD28739496
CAS Number: 1805936-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR