2-Bromo-6-fluoro-3-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30952-500MG 30952-1G 30952-5G 30952-25G
77325-178EA 140.08 USD
77325-178 77344-694 77361-516 77384-690
2-Bromo-6-fluoro-3-(methylthio)phenylboronic acid pinacol ester ≥97%
2-Bromo-6-fluoro-3-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088235
CAS Number: 2121512-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR