5-Fluoro-2-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30950-500MG 30950-1G 30950-5G 30950-25G
77325-176EA 101.97 USD
77325-176 77342-986 77361-514 77373-206
5-Fluoro-2-(methylthio)phenylboronic acid pinacol ester ≥97%
5-Fluoro-2-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088233
CAS Number: 1428532-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR