5-Bromo-2,3-difluoro-4-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30947-500MG 30947-1G 30947-5G 30947-25G
77325-174EA 134.93 USD
77325-174 77344-432 77361-512 77384-688
5-Bromo-2,3-difluoro-4-methylphenylboronic acid pinacol ester ≥97%
5-Bromo-2,3-difluoro-4-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BBrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29088232
CAS Number: 2121513-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR