5-Bromo-4-chloro-2,3-difluorophenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

30943-500MG 30943-1G 30943-5G
77337-044EA 407.88 USD
77337-044 77348-948 77373-204
5-Bromo-4-chloro-2,3-difluorophenylboronic acid pinacol ester ≥95%
5-Bromo-4-chloro-2,3-difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₃BBrClF₂O₂
MW: 353.395 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088229
CAS Number: 2121514-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR