Methyl 2-chloro-3,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

30941-500MG 30941-1G 30941-5G
77336-330EA 387.28 USD
77336-330 77348-946 77373-202
Methyl 2-chloro-3,4-difluorobenzoate ≥97%
Methyl 2-chloro-3,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18417404
CAS Number: 1261562-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR