2-Chloro-3,4-difluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30937-500MG 30937-1G 30937-5G
77338-950EA 535.6 USD
77338-950 77348-944 77373-200
2-Chloro-3,4-difluorophenylboronic acid pinacol ester ≥97%
2-Chloro-3,4-difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₂O₂
MW: 274.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088227
CAS Number: 2121515-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR