6-Bromo-2,4-difluoro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30933-500MG 30933-1G 30933-5G 30933-25G
77325-172EA 128.75 USD
77325-172 77344-234 77361-510 77373-198
6-Bromo-2,4-difluoro-3-methylbenzyl alcohol ≥97%
6-Bromo-2,4-difluoro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD29088224
CAS Number: 1879026-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR