2,4-Dichloro-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30880-500MG 30880-1G 30880-5G 30880-25G
77325-168EA 210.12 USD
77325-168 77348-940 77361-508 77384-686
2,4-Dichloro-5-methylphenylboronic acid pinacol ester ≥97%
2,4-Dichloro-5-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BCl₂O₂
MW: 286.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088221
CAS Number: 2121512-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR