3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(methylsulfonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

30874-500MG 30874-1G 30874-5G
77339-938EA 685.98 USD
77339-938 77348-938 77373-194
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(methylsulfonyl)pyridine ≥97%
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(methylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄S
MW: 297.178 g/mol
Storage Temperature: Ambient
CAS Number: 1445651-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR