2-Chloro-4-(methylthio)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
30841-1G
30841-5G
30841-25G
77340-870EA
61.8
USD
77340-870
77361-506
77373-192
2-Chloro-4-(methylthio)pyridine ≥97%
2-Chloro-4-(methylthio)pyridine
Precaución: for Research Use Only
Formula:
C₆H₆ClNS MW: 159.637 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07437890 CAS Number: 71506-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...