2,5-Difluoro-4-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2,5-Difluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

30840-500MG 30840-1G 30840-5G 30840-25G
77325-166EA 224.54 USD
77325-166 77348-936 77361-504 77384-684
2,5-Difluoro-4-methoxyphenylboronic acid pinacol ester ≥97%
2,5-Difluoro-4-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996271
CAS Number: 2121515-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR