2,4-Dibromo-5-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30838-500MG 30838-1G 30838-5G 30838-25G
77325-164EA 284.28 USD
77325-164 77348-934 77361-502 77384-682
2,4-Dibromo-5-fluorobenzyl alcohol ≥97%
2,4-Dibromo-5-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28784713
CAS Number: 1806354-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR