5-Chloro-2,6-dimethoxypyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

30826-500MG 30826-1G 30826-5G 30826-25G
77325-162EA 270.89 USD
77325-162 77348-932 77361-498 77384-680
5-Chloro-2,6-dimethoxypyridine-3-carboxylic acid ≥97%
5-Chloro-2,6-dimethoxypyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₄
MW: 217.606 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755887
CAS Number: 2755724-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR