3-Chloro-2-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

30795-500MG 30795-1G 30795-25G 30795-5G
77325-156EA 304.88 USD
77325-156 77348-926 77384-674 77373-186
3-Chloro-2-phenylpyridine ≥97%
3-Chloro-2-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈ClN
Storage Temperature: Ambient
MDL Number: MFCD10574690
CAS Number: 634198-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR