2-Chloro-3-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30783-1G 30783-5G 30783-25G
77341-394EA 84.46 USD
77341-394 77361-490 77373-184
2-Chloro-3-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
2-Chloro-3-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18756700
CAS Number: 1689550-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR