2,6-dimethoxy-3-(trimethylsilyl)pyridine-5-boronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
30762-500MG
30762-1G
30762-5G
30762-25G
77325-154EA
149.35
USD
77325-154
77345-298
77361-488
77384-672
2,6-dimethoxy-3-(trimethylsilyl)pyridine-5-boronic acid pinacol ester ≥97%
2,6-dimethoxy-3-(trimethylsilyl)pyridine-5-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₆H₂₈BNO₄Si MW: 337.294 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29088201 CAS Number: 2121513-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...