2,6-Dimethoxy-3-(trimethylsilyl)pyridine-5-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
30761-500MG
30761-1G
30761-5G
30761-25G
77325-152EA
98.88
USD
77325-152
77342-736
77361-486
77373-182
2,6-Dimethoxy-3-(trimethylsilyl)pyridine-5-boronic acid ≥97%
2,6-Dimethoxy-3-(trimethylsilyl)pyridine-5-boronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₈BNO₄Si MW: 255.151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29088200 CAS Number: 2121514-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...