2,6-Dimethoxy-5-methylpyridine-3-methanol ≥97%

Supplier: AOBChem USA

Warning

30756-500MG 30756-1G 30756-5G 30756-25G
77325-150EA 243.08 USD
77325-150 77348-924 77361-484 77384-670
2,6-Dimethoxy-5-methylpyridine-3-methanol ≥97%
2,6-Dimethoxy-5-methylpyridine-3-methanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD29088197
CAS Number: 1879026-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR