5-Bromo-2,6-dimethoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

30737-500MG 30737-1G 30737-5G 30737-25G
77325-144EA 160.68 USD
77325-144 77345-764 77361-478 77384-664
5-Bromo-2,6-dimethoxypyridin-3-ol ≥97%
5-Bromo-2,6-dimethoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₇H₈BrNO₃
MW: 234.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088186
CAS Number: 1826110-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR