N-Methoxy-N,1-dimethyl-1H-pyrazole-5-carboxamide ≥97%

Supplier: AOBChem USA

Warning

30703-500MG 30703-1G 30703-25G 30703-5G
77325-138EA 301.79 USD
77325-138 77348-918 77384-662 77373-176
N-Methoxy-N,1-dimethyl-1H-pyrazole-5-carboxamide ≥97%
N-Methoxy-N,1-dimethyl-1H-pyrazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18841493
CAS Number: 1339452-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR