2-Chloro-4-fluoro-6-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

30686-500MG 30686-1G 30686-5G 30686-25G
77325-136EA 216.3 USD
77325-136 77348-916 77361-472 77384-660
2-Chloro-4-fluoro-6-methylbenzoic acid ≥95%
2-Chloro-4-fluoro-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712110
CAS Number: 1427391-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR