2-(2,3-Dichloro-6-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

30679-500MG 30679-1G 30679-5G
77337-834EA 449.08 USD
77337-834 77348-912 77373-172
2-(2,3-Dichloro-6-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,3-Dichloro-6-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₆BCl₂FO₂
MW: 304.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088175
CAS Number: 2121513-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR